ENA408615752 Enamine

1-(2-bromo-5-fluoropyridin-4-yl)ethan-1-one

  • MDL number MFCD15526922
  • Cas Number : 1114523-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127642
Availability details for
Quantity
  • Availability
  • Add to Cart