ENA408615753 Enamine

3-bromo-5-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD18379742
  • Cas Number : 1261776-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136147
Availability details for
Quantity
  • Availability
  • Add to Cart