ENA408615758 Enamine

2-(4-amino-5-methyl-1H-pyrazol-1-yl)acetic acid

  • MDL number MFCD18307214
  • Cas Number : 1343304-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136166
Availability details for
Quantity
  • Availability
  • Add to Cart