ENA408615786 Enamine

N-(2-amino-2-methylpropyl)cyclopropanamine

  • MDL number MFCD14609229
  • Cas Number : 1247812-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136243

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136243
Availability details for
Quantity
  • Availability
  • Add to Cart