ENA408615790 Enamine

2-(ethylsulfanyl)ethane-1-thiol

  • MDL number MFCD00603035
  • Cas Number : 26750-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136247
Availability details for
Quantity
  • Availability
  • Add to Cart