ENA408615793 Enamine

1,3-dimethyl-5-(1H-1,2,4-triazol-1-yl)-1H-pyrazole-4-carboximidamide hydrochloride

  • MDL number MFCD26407708
  • Cas Number : 1461709-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136251
Availability details for
Quantity
  • Availability
  • Add to Cart