ENA408615797 Enamine

methyl 2-oxo-2H,3H-[1,3]oxazolo[4,5-b]pyridine-6-carboxylate

  • MDL number MFCD18074082
  • Cas Number : 1379222-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136258
Availability details for
Quantity
  • Availability
  • Add to Cart