ENA408615813 Enamine

{2-[(4-methoxyphenyl)methoxy]pyridin-4-yl}methanamine

  • MDL number MFCD11147546
  • Cas Number : 869293-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136329
Availability details for
Quantity
  • Availability
  • Add to Cart