ENA408615814 Enamine

N-Benzo[1,3]dioxol-5-ylmethyl-2-chloro-acetamide

  • MDL number MFCD02317455
  • Cas Number : 40023-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-01658

Safety Information

msds https://www.enaminestore.com/catalog/EN300-01658
Availability details for
Quantity
  • Availability
  • Add to Cart