ENA408615815 Enamine

2-[5-(1-methyl-1H-pyrazol-4-yl)-1,2,4-oxadiazol-3-yl]azepane

  • MDL number MFCD26407562
  • Cas Number : 1461705-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130285

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130285
Availability details for
Quantity
  • Availability
  • Add to Cart