ENA408615817 Enamine

1-(3-fluorophenyl)-2-(propan-2-yloxy)ethan-1-ol

  • MDL number MFCD21953124
  • Cas Number : 1461707-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136372
Availability details for
Quantity
  • Availability
  • Add to Cart