ENA408615818 Enamine

(2-cyclopropylethyl)(ethyl)amine

  • MDL number MFCD13249718
  • Cas Number : 760938-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136373

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136373
Availability details for
Quantity
  • Availability
  • Add to Cart