ENA408615819 Enamine

ethyl 4-(chlorosulfonyl)-1-methyl-1H-pyrrole-2-carboxylate

  • MDL number MFCD21224754
  • Cas Number : 868963-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136374

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136374
Availability details for
Quantity
  • Availability
  • Add to Cart