ENA408615820 Enamine

1-(3-cyclopropyl-1,2,4-oxadiazol-5-yl)ethan-1-ol

  • MDL number MFCD18335688
  • Cas Number : 1342181-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136375
Availability details for
Quantity
  • Availability
  • Add to Cart