ENA408615821 Enamine

1,2,3,4-tetrahydroquinoline-1-carbaldehyde

  • MDL number MFCD00024049
  • Cas Number : 2739-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136376
Availability details for
Quantity
  • Availability
  • Add to Cart