ENA408615825 Enamine

1-[1-phenyl-5-(trifluoromethyl)-1H-pyrazol-4-yl]methanamine

  • MDL number MFCD22223470
  • Cas Number : 1461707-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136381
Availability details for
Quantity
  • Availability
  • Add to Cart