ENA408615826 Enamine

5-amino-2,4-dimethylphenol

  • MDL number MFCD19203755
  • Cas Number : 14106-48-0
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136382
Availability details for
Quantity
  • Availability
  • Add to Cart