ENA408615832 Enamine

methyl 2-methyl-4-oxopentanoate

  • MDL number MFCD16068188
  • Cas Number : 32811-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136392
Availability details for
Quantity
  • Availability
  • Add to Cart