ENA408615841 Enamine

2-bromo-3,5-dimethylbenzoic acid

  • MDL number MFCD24108515
  • Cas Number : 7697-33-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136448

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136448
Availability details for
Quantity
  • Availability
  • Add to Cart