ENA408615843 Enamine

2-(1-methyl-1H-pyrazol-5-yl)ethan-1-amine

  • MDL number MFCD19620954
  • Cas Number : 807315-40-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136466

Safety Information

signalword Danger
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P310,P330,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-136466
pictograms GHS05,GHS07
hstatements H302,H318
Availability details for
Quantity
  • Availability
  • Add to Cart