ENA408615847 Enamine

5-methyl-6-phenyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one

  • MDL number MFCD21291634
  • Cas Number : 109942-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136470
Availability details for
Quantity
  • Availability
  • Add to Cart