ENA408615853 Enamine

2-[1-(azetidin-3-yl)-1H-1,2,3-triazol-4-yl]propan-2-ol hydrochloride

  • MDL number MFCD26407735
  • Cas Number : 1461709-27-2
  • Molecular Weight : 218.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-(azetidin-3-yl)-1H-1,2,3-triazol-4-yl)propan-2-ol hydrochloride
Inchi Code 1S/C8H14N4O.ClH/c1-8(2,13)7-5-12(11-10-7)6-3-9-4-6;/h5-6,9,13H,3-4H2,1-2H3;1H
Inchi Key PMOJVGMIGLHKGC-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136480

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136480
Availability details for
Quantity
  • Availability
  • Add to Cart