ENA408615854 Enamine

2-amino-2-methyl-3-(1H-1,2,4-triazol-1-yl)propanoic acid dihydrochloride

  • MDL number MFCD26407736
  • Cas Number : 1461706-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136482
Availability details for
Quantity
  • Availability
  • Add to Cart