ENA408615870 Enamine

2-methyl-3-(oxan-4-yl)propan-1-ol

  • MDL number MFCD20385991
  • Cas Number : 1343797-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131522
Availability details for
Quantity
  • Availability
  • Add to Cart