ENA408615871 Enamine

ethyl 2-(4-chloro-3-fluorophenyl)-2-hydroxyacetate

  • MDL number MFCD16694002
  • Cas Number : 1247619-04-0
  • Molecular Weight : 232.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136553
Availability details for
Quantity
  • Availability
  • Add to Cart