ENA408615874 Enamine

2-[2-(2-butoxyethoxy)ethoxy]ethane-1-sulfonyl chloride

  • MDL number MFCD19675992
  • Cas Number : 1343961-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136558
Availability details for
Quantity
  • Availability
  • Add to Cart