ENA408615889 Enamine

tert-butyl 4-chloro-3-nitrobenzoate

  • MDL number MFCD18452093
  • Cas Number : 157160-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-131760
Availability details for
Quantity
  • Availability
  • Add to Cart