ENA408615892 Enamine

3-fluoro-4-(propan-2-yl)aniline

  • MDL number MFCD21234657
  • Cas Number : 1369809-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136637
Availability details for
Quantity
  • Availability
  • Add to Cart