ENA408615895 Enamine

4-(2-chloroacetyl)benzonitrile

  • MDL number MFCD13172603
  • Cas Number : 40805-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136650
Availability details for
Quantity
  • Availability
  • Add to Cart