ENA408615899 Enamine

1,4-dimethyl-3-(trifluoromethyl)-1H-pyrazol-5-amine

  • MDL number MFCD11209554
  • Cas Number : 164668-13-7
  • Molecular Weight : 179.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 70-71
Iupac Name 1,4-dimethyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
Inchi Code 1S/C6H8F3N3/c1-3-4(6(7,8)9)11-12(2)5(3)10/h10H2,1-2H3
Inchi Key QRLFNOIUGQYYQU-UHFFFAOYSA-N

Description

Melting Point 70-71
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136656

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-136656
pictograms GHS05,GHS07
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart