ENA408615906 Enamine

2-[(2-chlorophenyl)methoxy]-5-methylbenzoic acid

  • MDL number MFCD13231162
  • Cas Number : 1053116-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136665
Availability details for
Quantity
  • Availability
  • Add to Cart