ENA408615914 Enamine

dimethyl-1H-1,2,3-triazol-4-amine

  • MDL number MFCD21947964
  • Cas Number : 1461713-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136684
Availability details for
Quantity
  • Availability
  • Add to Cart