ENA408615916 Enamine

3-(pentafluoro-lambda6-sulfanyl)aniline

  • MDL number MFCD00674199
  • Cas Number : 2993-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-37
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-136698

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P310,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes T,Xi
MSDS https://www.enaminestore.com/catalog/EN300-136698
pictograms GHS06
hstatements H301,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart