ENA408615922 Enamine

2,8-diazaspiro[4.5]decane

  • MDL number MFCD16877059
  • Cas Number : 176-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136719
Availability details for
Quantity
  • Availability
  • Add to Cart