ENA408615923 Enamine

3,5-dimethyl-4-[(pyrrolidin-3-yloxy)methyl]-1,2-oxazole

  • MDL number MFCD16709384
  • Cas Number : 1248491-43-1
  • Molecular Weight : 196.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dimethyl-4-[(3-pyrrolidinyloxy)methyl]isoxazole
Inchi Code 1S/C10H16N2O2/c1-7-10(8(2)14-12-7)6-13-9-3-4-11-5-9/h9,11H,3-6H2,1-2H3
Inchi Key LXZZSHUNASWVTN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136720
Availability details for
Quantity
  • Availability
  • Add to Cart