ENA408615928 Enamine

ethyl 2-methyl-3-(4-nitrophenyl)prop-2-enoate

  • Cas Number : 53618-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131875
Availability details for
Quantity
  • Availability
  • Add to Cart