ENA408615929 Enamine

5-bromo-4-methyl-2-nitropyridine

  • MDL number MFCD17014995
  • Cas Number : 1225278-70-5
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-methyl-2-nitropyridine
Inchi Code 1S/C6H5BrN2O2/c1-4-2-6(9(10)11)8-3-5(4)7/h2-3H,1H3
Inchi Key OZAONQSRAXEGMW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136734

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-136734
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart