ENA408615931 Enamine

7-(3-aminopropyl)-5,7-diazaspiro[3.4]octane-6,8-dione

  • MDL number MFCD21909216
  • Cas Number : 1368587-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131893
Availability details for
Quantity
  • Availability
  • Add to Cart