ENA408615943 Enamine

(2-phenylbutanoyl)urea

  • MDL number MFCD00069050
  • Cas Number : 90-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136805
Availability details for
Quantity
  • Availability
  • Add to Cart