ENA408615946 Enamine

[3-(propan-2-yl)-1,2,4-thiadiazol-5-yl]methanol

  • MDL number MFCD18804525
  • Cas Number : 337904-88-8
  • Molecular Weight : 158.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-isopropyl-1,2,4-thiadiazol-5-yl)methanol
Inchi Code 1S/C6H10N2OS/c1-4(2)6-7-5(3-9)10-8-6/h4,9H,3H2,1-2H3
Inchi Key FZEQFDKOVBEFJR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136808
Availability details for
Quantity
  • Availability
  • Add to Cart