ENA408615949 Enamine

pyridin-4-ylmethanethiol

  • MDL number MFCD06657437
  • Cas Number : 1822-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136811

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-136811
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart