ENA408615950 Enamine

7-tert-butyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD20282731
  • Cas Number : 57817-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136812
Availability details for
Quantity
  • Availability
  • Add to Cart