ENA408615952 Enamine

4-cyclobutoxypyridine-2-carbonitrile

  • MDL number MFCD17252744
  • Cas Number : 1344306-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132267

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132267
Availability details for
Quantity
  • Availability
  • Add to Cart