ENA408615965 Enamine

5-[(4-chlorophenyl)sulfanyl]-2-nitrobenzoic acid

  • MDL number MFCD13352202
  • Cas Number : 1036467-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136856
Availability details for
Quantity
  • Availability
  • Add to Cart