ENA408615968 Enamine

3-{[(1H-pyrazol-4-yl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD12902725
  • Cas Number : 1183510-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136862
Availability details for
Quantity
  • Availability
  • Add to Cart