ENA408615973 Enamine

4-(4-bromo-1,3-thiazol-2-yl)pyridine

  • MDL number MFCD11869690
  • Cas Number : 1142195-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136873
Availability details for
Quantity
  • Availability
  • Add to Cart