ENA408615974 Enamine

2-(2-chloro-6-fluorophenyl)-2-(N-methylacetamido)acetic acid

  • MDL number MFCD19175585
  • Cas Number : 1270614-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136880
Availability details for
Quantity
  • Availability
  • Add to Cart