ENA408615986 Enamine

methyl 2-(benzyloxy)-5-chloro-4-methylbenzoate

  • MDL number MFCD26407587
  • Cas Number : 1461708-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-132465
Availability details for
Quantity
  • Availability
  • Add to Cart