ENA408615994 Enamine

4-(3-hydroxypropoxy)-3-methoxybenzonitrile

  • MDL number MFCD12179515
  • Cas Number : 1157110-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132556

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132556
Availability details for
Quantity
  • Availability
  • Add to Cart