ENA408616001 Enamine

5-(pyridin-4-yl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD01692465
  • Cas Number : 3652-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 271-276
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-02625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-02625
Availability details for
Quantity
  • Availability
  • Add to Cart